missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AS160 Monoclonal Antibody (G.853.7)

Rabbit Monoclonal Antibody
Marca: Invitrogen™ MA514840
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
It is not recommended to aliquot this antibody.
AS160 (TBC1D4) is a GTPase-activiting protein for RAB2A, RAB8A, RAB10, and RAB14. The isoform 2 version of this protein promotes the insulin-induced glucose transporter SLC2A4/GLUT4 translocation at the plasma membrane. This movement increases glucose uptake. Mutations associated with the gene cause Diabetes mellitus, non-insulin-dependent, 5 (NIDDM5).
Specifica
| AS160 | |
| Monoclonal | |
| 81 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| O60343, Q8BYJ6 | |
| TBC1D4 | |
| Synthetic peptide corresponding to residues surrounding Ala195 of human AS160. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| G.853.7 | |
| Unconjugated | |
| TBC1D4 | |
| 5930406J04Rik; A930035N22; Akt substrate of 160 kDa; AS160; AV295684; DKFZp779C0666; Kiaa0603; NIDDM5; OTTHUMP00000040818; RGD1561609; RP11-159J2.1; RP11-159J2.3; similar to TBC1 domain family member 4; TBC (Tre-2, BUB2, CDC16) domain-containing protein; TBC Tre-2; TBC1 domain family member 4; TBC1 domain family, member 4; TBC1D4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 210789, 306117, 9882 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto