missing translation for 'onlineSavingsMsg'
Learn More

β-Arrestin Mouse, Unlabeled, Clone: 10, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610551

 View more versions of this product

Product Code. 15815519

  • ONLINE EXCLUSIVE PRICE
    £419.00
    £401.97 / 1 set
    Save £17.03
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

β-Arrestins were discovered due to their ability to modulate interactions between the phosphorylated β2-Adrenergic receptors and G proteins. This modulation results in diminished β2-Adrenergic receptor function, also known as desensitization. Because arrestins are found at the synaptic terminals, they may provide a termination mechanism that allows the neurons to regain their original polarization and respond to a new neurotransmitter stimulus. The C-terminal region of arrestins is involved in selecting the phosphorylated and activated adrenergic receptors. The β-Arrestin1 gene encodes a protein of 418 amino acids with an approximate molecular weight of 55kDa. β-Arrestin1 protein is highly homologous to the 45kDa β-Arrestin2. Both proteins are widely expressed, but are especially abundant in the central nervous system.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

β-Arrestin
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Rat β-Arrestin1 aa. 262-409
150 μg
Cell Biology
Human, Mouse, Rat
IgG1
Western Blot
10
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.