Learn More
actin related protein 2/3 complex, subunit 5, 16kDa, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant ARPC5.
Brand: Abnova H00010092-A01.50uL
Description
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined. [provided by RefSeq
Sequence: MSKNTVSSARFRKVDVDEYDENKFVDEEDGGDGQAGPDEGEVDSCLRHSITGNMTAALQAALKNPPINTKSQAVKDRAGSIVLKVLISFKANDIEKAVQSLDKNGVDLLMKYIYKGFESPSDNSSAMLLQWHEKALAAGGVGSIVRVLTARKTVSpecifications
| actin related protein 2/3 complex, subunit 5, 16kDa | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant ARPC5. | |
| ARPC5 | |
| ARC16/MGC88523/dJ127C7.3/p16-Arc | |
| Mouse | |
| 50 μL | |
| Yes | |
| 10092 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC057237 | |
| ARPC5 | |
| ARPC5 (AAH57237, 1 a.a. ∼ 154 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.