missing translation for 'onlineSavingsMsg'
Learn More

Rho guanine nucleotide exchange factor (GEF) 5, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ARHGEF5 protein.

Brand:  Abnova H00007984-B01.50uL

Product Code. 16158105

  • £303.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Rho GTPases play a fundamental role in numerous cellular processes initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein may be involved in the control of cytoskeletal organization. [provided by RefSeq

Sequence: MGGFSRRCSKLINSSQLLYQEYSDVVLNKEIQSQQRLESLSETPGPSSPRQPRKALVSSESYLQRLSMASSGSLWQEIPVVRNSTVLLSMTHEDQKLQEVKFELIVSEASYLRSLNIAVDHFQLSTSLRATLSNQEHQWLFSRLQDVRDVSATFLSDLEENFENNIFSFQVCDVVLNHAPDFRRVYLPYVTNQTYQERTFQSLMNSNSNFREVLEKLESDPVCQRLSLKSFLILPFQRITRLKLLLQNILKRTQPGSSEEAEATKAHHALEQLIRDCNNNVQSMRRTEELIYLSQKIEFECKIFPLISQSRWLVKSGELTALEFSASPGLRRKLNTRPVHLHLFNDCLLLSRPREGSRFLVFDHAPFSSIRGEKCEMKLHGPHKNLFRLFLRQNTQGAQAEFLFRTETQSEKLRWISALAMPREELDLLECYNSPQVQCLRAYKPRENDELALEKADVVMVTQQSSDGWLEGVRLSDGERGWFPVQQVEFISNPEVRAQNLKEAHRVKTAKLQLVEQQA
Specifications

Specifications

Rho guanine nucleotide exchange factor (GEF) 5
Polyclonal
Mouse polyclonal antibody raised against a full-length human ARHGEF5 protein.
ARHGEF5
DKFZp686N1969/GEF5/P60/TIM/TIM1
Mouse
50 μL
Yes
7984
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Western Blot
Unconjugated
No additive
Q12774
ARHGEF5
ARHGEF5 (Q12774, 1 a.a. ∼ 519 a.a) full-length human protein.
RUO
Primary
Human
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.