missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518201
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. This protein is similar to rat collybistin protein. Alternative splicing of this gene generates two transcript variants which encode different isoforms. Also there is possibility for the usage of multiple polyadenylation sites for this gene.Specifications
ARHGEF4 | |
Polyclonal | |
Unconjugated | |
Arhgef4 | |
9330140K16Rik; APC-stimulated guanine nucleotide exchange factor; APC-stimulated guanine nucleotide exchange factor 1; Arhgef4; Asef; ASEF1; DKFZp434G2016; ENSMUSG00000070955; GEF4; Kiaa1112; LOC100067370; LOW QUALITY PROTEIN: rho guanine nucleotide exchange factor 4; Rho guanine nucleotide exchange factor (GEF) 4; Rho guanine nucleotide exchange factor 4; STM6 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
50649 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9NR80 | |
Arhgef4 | |
Synthetic peptide sequence (PWEEPAGEKPSCS) corresponding to the N-terminus amino acids of ARHGEF4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |