Learn More
Rho GDP dissociation inhibitor (GDI) alpha, Mouse, Clone: 1E12-2G11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ARHGDIA.
Brand: Abnova H00000396-M01.100ug
Description
Aplysia Ras-related homologs (ARHs), also called Rho genes, belong to the RAS gene superfamily encoding small guanine nucleotide exchange (GTP/GDP) factors. The ARH proteins may be kept in the inactive, GDP-bound state by interaction with GDP dissociation inhibitors, such as ARHGDIA (Leffers et al., 1993 [PubMed 8262133]).[supplied by OMIM
Sequence: MAEQEPTAEQLAQIAAENEEDEHSVNYKPPAQKSIQEIQELDKDDESLRKYKEALLGRVAVSADPNVPNVVVTGLTLVCSSAPGPLELDLTGDLESFKKQSFVLKEGVEYRIKISFRVNREIVSGMKYIQHTYRKGVKIDKTDYMVGSYGPRAEEYEFLTPVEEAPKGMLARGSYSIKSRFTDDDKTDHLSWEWNLTIKKDWKDSpecifications
| Rho GDP dissociation inhibitor (GDI) alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC016031 | |
| ARHGDIA | |
| ARHGDIA (AAH16031, 1 a.a. ∼ 204 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Signal Transduction | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1E12-2G11 | |
| Mouse monoclonal antibody raised against a full length recombinant ARHGDIA. | |
| ARHGDIA | |
| GDIA1/MGC117248/RHOGDI/RHOGDI-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 396 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.