Learn More
ARHGAP4, Mouse, Clone: 3F7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ARHGAP4.
Brand: Abnova H00000393-M01.100ug
Description
This gene encodes a member of the rhoGAP family of proteins which play a role in the regulation of small GTP-binding proteins belonging to the RAS superfamily. The protein encoded by the orthologous gene in rat is localized to the Golgi complex and can redistribute to microtubules. The rat protein stimulates the activity of some Rho GTPases in vitro. Genomic deletions of this gene and a neighboring gene have been found in patients with nephrogenic diabetes insipidus. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: TSPEAMGPSGHRRRCLVPASPEQHVEVDKAVAQNMDSVFKELLGKTSVRQGLGPASTTSPSPGPRSPKAPPSSRLGRNKGFSRGPGAPASPSASHPQGLDTTPKPHSpecifications
| ARHGAP4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC052303 | |
| ARHGAP4 | |
| ARHGAP4 (AAH52303, 881 a.a. ∼ 986 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 3F7 | |
| Mouse monoclonal antibody raised against a partial recombinant ARHGAP4. | |
| ARHGAP4 | |
| C1/KIAA0131/RGC1/RhoGAP4/p115 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 393 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.