Learn More
anti-arginase I, Polyclonal PA522009, Invitrogen™
Specifically detect arginase I in Human, Mouse samples using Thermo Scientifc™ Pierce™ anti-arginase I Polyclonal, validated for ICC, IHC (P) and WB
Brand: Thermo Scientific PA5-22009
Beschreibung
Specifically detect arginase I in Human, Mouse samples using Thermo Scientifc™ Pierce™ anti-arginase I Polyclonal, validated for ICC, IHC (P) and WB
Host Species: Rabbit
Cross Reactivity: Human, Mouse
Isotype: IgG
Immunogen: Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 322 (P05089) of Arginase I
Formulation: 100μL of Antigen affinity chromatography purified antibody (1mg/mL) in PBS buffer, with glycerol 20%; pH 7.
Uniprot Accession: P05089, Q61176
Entrez Gene ID: 383, 11846
PA5-22009 detects arginase I in Human and Mouse samples. PA5-22009 has been successfully used in ICC, IHC (P) and WB procedures. The immunogen is Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 322 (P05089) of Arginase I.
Spezifikation
| arginase I | |
| Unconjugated | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG |
| Polyclonal | |
| ARG1 | |
| Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 272 (P05089) of Arginase I | |
| 100 μg | |
| Primary | |
| Store at -20°C. Avoid freeze/Thaw cycles. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.