Learn More
ADP-ribosylation factor 5, Mouse, Clone: 1B4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ARF5.
Brand: Abnova H00000381-M01.100ug
Description
ADP-ribosylation factor 5 (ARF5) is a member of the human ARF gene family. These genes encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking and as activators of phospholipase D. The gene products include 6 ARF proteins and 11 ARF-like proteins and constitute 1 family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2,and ARF3), class II (ARF4 and ARF5) and class III (ARF6). The members of each class share a common gene organization. The ARF5 gene spans approximately 3.2kb of genomic DNA and contains six exons and five introns. [provided by RefSeq
Sequence: YFQNTQGLIFVVDSNDRERVQESADELQKMLQEDELRDAVLLVFANKQDMPNAMPVSELTDKLGLQHLRSRTWYVQATCATQGTGLYDGLDWLSHELSKRSpecifications
| ADP-ribosylation factor 5 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC003043 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 1B4 | |
| Mouse monoclonal antibody raised against a partial recombinant ARF5. | |
| ARF5 | |
| ARF5 | |
| ARF5 (AAH03043, 81 a.a. ∼ 180 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Signal Transduction | |
| 381 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.