missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AIP Monoclonal Antibody (35-2)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA316515
This item is not returnable.
View return policy
Description
This antibody is specific for the FKBP domain. Suggested positive control: antigen standard for AIP (transient overexpression lysate).
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.
Specifications
| AIP | |
| Monoclonal | |
| Conc. Not Determined | |
| Tissue culture supernatant with 0.1% sodium azide | |
| O00170, O08915, Q5FWY5 | |
| Aip | |
| Bacterially expressed human AIP/ARA9. | |
| RUO | |
| 11632, 282827, 9049 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 35-2 | |
| Unconjugated | |
| Aip | |
| AA408703; AH receptor-interacting protein; AIP; ARA9; aryl hydrocarbon receptor interacting protein; aryl-hydrocarbon receptor-interacting protein; AW476050; D19Bwg1412e; FKBP16; FKBP37; HBV X-associated protein 2; HBV-X associated protein; immunophilin homolog ARA9; Immunophilin XAP2; SMTPHN; XAP2; XAP-2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering