missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHROOM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527090
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene shares significant similarities with the apical protein from Xenopus laevis which is implicated in amiloride-sensitive sodium channel activity. This gene is a strong candidate gene for ocular albinism type 1 syndrome.
Specifications
| SHROOM2 | |
| Polyclonal | |
| Unconjugated | |
| SHROOM2 | |
| 4832440C16; Ab2-404; apical-like protein; APX homolog of Xenopus; Apxl; C630003H05Rik; HSAPXL; liver regeneration-related protein LRRG167; protein Apxl; Protein Shroom2; Shrm2; shroom family member 2; SHROOM2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110380, 357 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| A2ALU4, Q13796 | |
| SHROOM2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human APXL. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction