missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APOE Monoclonal Antibody (4E4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518068
This item is not returnable.
View return policy
Description
This antibody is specific for Apolipoprotein E4 and shows no cross-reactivity with ApoE2 or ApoE3.
Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13. 2.
Specifications
| APOE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P02649 | |
| APOE | |
| Synthetic peptide corresponding to Human Apolipoprotein E4 conjugated to Keyhole Limpet Haemocyanin (KLH) (Immunogen sequence: ApoE4 peptide). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Dot Blot, ELISA, Immunoprecipitation, Western Blot, Western Blot | |
| 4E4 | |
| Unconjugated | |
| APOE | |
| AD2; AI255918; APOE; apo-E; ApoE4; APOEA; Apolipoprotein; apolipoprotein E; apolipoprotein E3; apolipoprotein E4; Apoprotein; B2G1; BG; LDLCQ5; LPG; MGC1571 | |
| Mouse | |
| Protein G | |
| RUO | |
| 348 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction