Learn More
APOC1, Mouse, Clone: 2E2-1A3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant APOC1.
Brand: Abnova H00000341-M01.100ug
Description
The protein encoded by this gene is a member of the apolipoprotein C1 family. This gene is expressed primarily in the liver, and it is activated when monocytes differentiate into macrophages. A pseudogene of this gene is located 4kb downstream in the same orientation, on the same chromosome. This gene is mapped to chromosome 19, where it resides within a apolipoprotein gene cluster. Alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined. [provided by RefSeq
Sequence: MRLFLSLPVLVVVLSIVLEGPAPAQGTPDVSSALDKLKEFGNTLEDKARELISRIKQSELSAKMREWFSETFQKVKEKLKIDSSpecifications
| APOC1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC009698 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 341 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Western Blot | |
| 2E2-1A3 | |
| Mouse monoclonal antibody raised against a full length recombinant APOC1. | |
| APOC1 | |
| APOC1 | |
| APOC1 (AAH09698, 1 a.a. ∼ 83 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.