Learn More
annexin A9, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ANXA9 protein.
Brand: Abnova H00008416-B01P.50ug
Description
The annexins are a family of calcium-dependent phospholipid-binding proteins. Members of the annexin family contain 4 internal repeat domains, each of which includes a type II calcium-binding site. The calcium-binding sites are required for annexins to aggregate and cooperatively bind anionic phospholipids and extracellular matrix proteins. This gene encodes a divergent member of the annexin protein family in which all four homologous type II calcium-binding sites in the conserved tetrad core contain amino acid substitutions that ablate their function. However, structural analysis suggests that the conserved putative ion channel formed by the tetrad core is intact. [provided by RefSeq
Sequence: MAPSLTQEILSHLGLASKTAAWGTLGTLRTFLNFSVDKDAQRLLRAITGQGVDRSAIVDVLTNRSREQRQLISRNFQERTQQDLMKSLQAALSGNLERIVMALLQPTAQFDAQELRTALKASDSAVDVAIEILATRTPPQLQECLAVYKHNFQVEAVDDITSETSGILQDLLLALAKGGRDSYSGIIDYNLAEQDVQALQRAEGPSREETWVPVFTQRNPEHLIRVFDQYQRSTGQELEEAVQNRFHGDAQVALLGLASVIKNTPLYFADKLHQALQETEPNYQVLIRILISRCETDLLSIRAEFRKKFGKSLYSSLQDAVKGDCQSALLALCRAEDMSpecifications
| annexin A9 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ANXA9 protein. | |
| ANXA9 | |
| ANX31 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 8416 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC005830.2 | |
| ANXA9 | |
| ANXA9 (AAH05830.2, 1 a.a. ∼ 338 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.