Learn More
anti-ANXA10 Matched Antibody Pair, Abnova
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-ANXA10 (100μg); Mouse monoclonal anti-ANXA10, IgG2bκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The function of this gene has not yet been determined.
ELISA
Specifications
Specifications
| Antigen | ANXA10 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ANXA10. |
| Gene | ANXA10 |
| Gene Alias | ANX14 |
| Gene Symbols | ANXA10 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.