Learn More
Annexin V anti-Human, PerCP-Cy5.5, Clone: , BD
Monoclonal Antibody
Brand: BD Biosciences 561431
Description
Apoptosis is a normal physiologic process which occurs during embryonic development as well as in maintenence of tissue homeostasis. The apoptotic program is characterized by certain morphologic features, including loss of plasma membrane asymmetry and attachment, condensation of the cytoplasm and nucleus, and internucleosomal cleavage of DNA. Loss of plasma membrane is one of the earliest features. In apoptotic cells, the membrane phospholipid phosphatidylserine (PS) is translocated from the inner to the outer leaflet of the plasma membrane, thereby exposing PS to the external cellular environment. Annexin V is a 35-36 kDa Ca2+ dependent phospholipid-binding protein that has a high affinity for PS, and binds to cells with exposed PS. Annexin V may be conjugated to fluorochromes including PerCP-Cy™5.5 Annexin V. This format retains its high affinity for PS and thus serves as a sensitive probe for flow cytometric analysis of cells that are undergoing apoptosis. Since externalization of PS occurs in the earlier stages of apoptosis, PerCP-Cy™5.5 Annexin V staining can identify apoptosis at an earlier stage than assays based on nuclear changes such as DNA fragmentation.
NOTE:Investigators should note that the use of Propidium Iodide (PI) or 7-Amino-Actinomycin D (7-AAD) for co-staining with PerCP-Cy5.5 Annexin Vis not recommended due to spectral overlap.
Specifications
| Annexin V | |
| Monoclonal | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| PerCP-Cyanine5.5 | |
| Affinity Purified | |
| RUO | |
| Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.