missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523954
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, mouse and porcine based on sequence homology.
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.Specifications
Angiopoietin 1 | |
Polyclonal | |
Unconjugated | |
Angpt1 | |
1110046O21Rik; 2810039D03Rik; AGP1; Agpt; Agpt1; ANG1; ang-1; ANG3; ANG-3; angioarrestin; angiopoietin 1; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-1; Angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; Angpt1; ANGPT3; Angptl1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); endothelial growth factor; KIAA0003; PSEC0154; tie2 receptor ligand; UNQ162; UNQ162/PRO188 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
284 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 0.09% sodium azide | |
Q15389 | |
Angpt1 | |
KLH conjugated synthetic peptide between 374-402 amino acids from the C-terminal region of human ANGPT1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |