missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Amphiphysin Polyclonal Antibody

Description
Recommended positive controls: mouse hippocampus, mouse brain, rat brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (91%), Pig (100%), Chicken (95%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Amphiphysins, members of the BAR (Bin-Amphiphysin-Rvsp) protein super family, play a key role in clathrin-mediated endocytosis of synaptic vesicles (SVs). All members of the BAR family share a highly conserved N-terminal BAR domain and a C-terminal Src homology (SH3) domain. Two isoforms of amphiphysin have been identified and act in concert as a heterodimer. Amphiphysins interact via its carboxy-terminal SH3 domain with dynamin, synaptojanin and clathrin. These complexes are implicated in synaptic vesicle recycling at nerve terminal. Amphiphysins were also identified as the dominant autoantigen in paraneoplastic Stiff-Man Syndrome.
Specifications
Specifications
| Antigen | Amphiphysin |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Amph |
| Gene Accession No. | O08838, P49418, Q7TQF7 |
| Gene Alias | Amph; AMPH1; amphiphysin; amphiphysin (Stiff-Mann syndrome with breast cancer 128kD autoantigen); amphiphysin 1; amphiphysin I; HGNC:471 |
| Gene Symbols | Amph |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?