missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPD2 (M09A), Mouse anti-Human, Clone: 6A8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant AMPD2.
Brand: Abnova H00000271-M09A.200uL
This item is not returnable.
View return policy
Description
Adenosine monophosphate deaminase-2 (EC 3.5.4.6) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle.[supplied by OMIM
Sequence: ISQDVKLEPDILLRAKQDFLKTDSDSDLQLYKEQGEGQGDRSLRERDVLEREFQRVTISGEEKCGVPFTDLLDAAKSVVRALFIREKYMALSLQSFCPTTSpecifications
| AMPD2 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_139156 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 6A8 | |
| Mouse monoclonal antibody raised against a partial recombinant AMPD2. | |
| AMPD2 | |
| AMPD2 | |
| AMPD2 (NP_631895, 86 a.a. ∼ 185 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 271 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction