missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMACR Monoclonal Antibody (R.16.1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA514793
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
P504S/AMACR/a-Methylacyl-CoA Racemase is an essential enzyme in the b-oxidation of branched-chain fatty acids. High expression of P504S protein is found in prostatic adenocarcinoma but not in benign prostate tissue by immunohistochemical staining in paraffin-embedded tissue. The expression of P504S is also detected in two premalignant lesions of the prostate: high-grade prostatic intraepithelial neoplasia (PIN) and atypical adenomatous hyperplasia.
Specifications
| AMACR | |
| Monoclonal | |
| Unconjugated | |
| AMACR | |
| 2-arylpropionyl-CoA epimerase; 2-methylacyl-CoA racemase; Alpha-methylacyl-CoA racemase; alpha-methylacyl-Coenzyme A racemase; Amacr; AMACRD; amcr; CBAS4; Da1-8; Macr1; Marc1; OTTHUMP00000219978; OTTHUMP00000219979; RACE; RM | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 17117, 23600 | |
| -20°C | |
| Liquid |
| Western Blot | |
| R.16.1 | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| O09174, Q9UHK6 | |
| AMACR | |
| Purified truncated recombinant human AMACR-His fusion protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction