Learn More
alpha Synuclein Recombinant Rabbit Monoclonal Antibody (14H2L1), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 701085
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
alpha Synuclein Recombinant Monoclonal antibody specifically detects alpha Synuclein in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
alpha Synuclein | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor) | |
Peptide corresponding to amino acids 117-125 of human synuclein, alpha | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
14H2L1 | |
Unconjugated | |
P37840 | |
Rabbit | |
Protein A | |
RUO | |
6622 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |