missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sarcomeric alpha Actinin Monoclonal Antibody (EA-53)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA122863
This item is not returnable.
View return policy
Description
MA1-22863 detects sarcomeric alpha Actinin in bovine, feline, chicken, fish, goat, hamster, lizard, mouse porcine, rabbit, rat, ovine, snake, and Xenopus laevis samples.
This antibody is specific for alpha-skeletal muscle actinin and alpha-cardiac actinin. It stains Z lines and dots in stress fibers of myotubes in skeletal and cardiac muscle but not in non-sarcomeric muscle elements.
Specifications
| Sarcomeric alpha Actinin | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.09% sodium azide | |
| P12814, Q7TPR4, Q9Z1P2 | |
| ACTN1 | |
| Purified rabbit skeletal alpha-actinin. | |
| RUO | |
| 100341378, 109711, 560400, 81634, 87 | |
| 4°C | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| EA-53 | |
| Unconjugated | |
| ACTN1 | |
| 3110023F10Rik; actinin 1 smooth muscle; actinin alpha 1; actinin, alpha 1; ACTN1; Actn1a; alpha actinin 1a; alpha-actinin cytoskeletal isoform; Alpha-actinin-1; BDPLT15; F-actin cross-linking protein; non-muscle alpha-actinin 1; non-muscle alpha-actinin-1 | |
| Mouse | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Zebrafish | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction