Learn More
aldehyde dehydrogenase 6 family, member A1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ALDH6A1 protein.
Brand: Abnova H00004329-B01.50uL
Description
This protein belongs to the aldehyde dehydrogenases family of proteins. This enzyme plays a role in the valine and pyrimidine catabolic pathways. The product of this gene, a mitochondrial methylmalonate semialdehyde dehydrogenase, catalyzes the irreversible oxidative decarboxylation of malonate and methylmalonate semialdehydes to acetyl- and propionyl-CoA. Methylmalonate semialdehyde dehydrogenase deficiency is characterized by elevated beta-alanine, 3-hydroxypropionic acid, and both isomers of 3-amino and 3-hydroxyisobutyric acids in urine organic acids. [provided by RefSeq
Sequence: MAALLAAAAVRARILQVSSKVKSSPTWYSASSFSSSVPTVKLFIGGKFVESKSDKWIDIHNPATNEVIGRVPQATKAEMDAAIASCKRAFPAWADTSVLSRQQVLLRYQQLIKENLKEIAKLITLEQGKTLADAEGDVFRGLQVVEHACSVTSLMMGETMPSITKDMDLYSYRLPLGVCAGIAPFNFPAMIPLWMFPMAMVCGNTFLMKPSERVPGATMLLAKLLQDSGAPDGTLNIIHGQHEAVNFICDHPDIKAISFVGSNKAGEYIFERGSRHGKRVQANMGAKNHGVVMPDANKENTLNQLVGAAFGAAGQRCMALSTAVLVGEAKKWLPELVEHAKNLRVNAGDQPGADLGPLITPQAKERVCNLIDSGTKEGASILLDGRKIKVKGYENGNFVGPTIISNVKPNMTCYKEEIFGPVLVVLETETLDEAIQIVNNNPYGNGTAIFTTNGATARKYAHLVDVGQVGVNVPIPVPLPMFSFTGSRSSFRGDTNFYGKQGIQFYTQLKTITSQWKEEDATLSSPAVVMPTMGRSpecifications
| aldehyde dehydrogenase 6 family, member A1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ALDH6A1 protein. | |
| ALDH6A1 | |
| MGC40271/MMSADHA/MMSDH | |
| Mouse | |
| 50 μL | |
| Yes | |
| 4329 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_005589.2 | |
| ALDH6A1 | |
| ALDH6A1 (NP_005580.1, 1 a.a. ∼ 535 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.