Learn More
ALDH Mouse anti-Human, Unlabeled, Clone: 44, BD
Mouse Monoclonal Antibody
£163.84 - £312.61
Specifications
| Antigen | ALDH |
|---|---|
| Clone | 44 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15895829
|
BD Biosciences
611194 |
50 μg |
£171.00 £163.84 / 1 set Save £7.16 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15805839
|
BD Biosciences
611195 |
150 μg |
£326.00 £312.61 / 1 set Save £13.39 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aldehyde dehydrogenase (ALDH) is a ubiquitous enzyme located in nearly all mammalian tissues. It catalyzes the irreversible oxidation of a range of aliphatic and aromatic aldehydes to their corresponding carboxylic acids. There are multiple isoforms of ALDH which are subdivided into three classes. Class I includes the cytosolic isoforms. Class II includes the mitochondrial isoforms. Class III includes the microsomal, cytosolic tumor specific, and cytosolic dioxin-inducible forms. At least twelve human ALDH isoforms have been identified. Mutations of many of these proteins such as ALDH1, ALDH2, ALDH4, and ALDH10 have been implicated in multiple human metabolic disorders and clinical abnormalities. At the amino acid level, human ALDH isoforms exhibit a wide range of diversity (15% to about 80%). However, multiple protein regions have been highly conserved and are important for functional activities. A well-characterized member of the human ALDH family is ALDH1. It plays a major role in the biosynthesis of retinoic acid from retinol (vitamin A). Retinoic acid, the biologically
active form of retinol, is a regulator of cellular proliferation, differentiation, and survival.
Host Species: Mouse
Clone: 44
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human ALDH1 aa. 7-128
Formula Weight [Chemical]: 55kDa
Immunofluorescence, Western Blotting
Specifications
| ALDH | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 44 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Human ALDH1 aa. 7-128 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.