Learn More
AKAP9 Monoclonal Antibody (17G10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1803
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-803 detects the AKAP450 in human and mouse cells. MA1-803 has been successfully used in Western blot, immunoprecipitation, immunohistochemistry, and immunofluorescence procedures. The MA1-803 immunogen is a synthetic peptide corresponding to residues Q(31) F R Q R K A Q S D G Q S P S(45) of human AKAP450.
AKAP9 Monoclonal antibody specifically detects AKAP9 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
AKAP9 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q70FJ1, Q99996 | |
AKAP9 | |
Synthetic peptide corresponding to residues Q(31) F R Q R K A Q S D G Q S P S(45) of human AKAP450. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
17G10 | |
Unconjugated | |
AKAP9 | |
5730481H23Rik; A kinase (PRKA) anchor protein (yotiao) 9; A kinase (PRKA) anchor protein 9; AKAP 120-like protein; AKAP 350; AKAP 450; AKAP350; AKAP450; AKAP9; AKAP-9; AKAP9-BRAF fusion protein; A-kinase anchor protein 350 kDa; A-kinase anchor protein 450 kDa; A-kinase anchor protein 9; A-kinase anchoring protein 9; AW545847; C79026; centrosome- and Golgi-localized PKN-associated protein; centrosome- and golgi-localized protein kinase N-associated protein; CG-NAP; G1-448-15; HYPERION; KIAA0803; kinase N-associated protein; LQT11; mei2-5; mKIAA0803; MU-RMS-40.16A; PPP1R45; PRKA9; Protein hyperion; protein kinase A anchoring protein 9; protein kinase A-anchoring protein 9; protein phosphatase 1, regulatory subunit 45; protein yotiao; repro12; YOTIAO | |
Mouse | |
Protein G | |
RUO | |
100986, 10142 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |