missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKAP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536155
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 47 and 79 kDa. Purity is >95% by SDS-PAGE.
AKAPs (A-kinase anchoring proteins), as their name implies, are a family of scaffolding proteins that bind regulatory subunits of Protein Kinase A (PKA) thus localizing PKA activity to distinct regions of the cell. Beyond a common amphipathic alpha helix that is responsible for recruiting the PKA regulatory subunit (RI alpha, RII alpha, RI beta, or RII beta), individual AKAPs contain additional domains responsible for the recrutiment of additional signaling proteins (phosphodiesterases, phosphatases, cytoskeletal components, other kinase, etc.) or restricting AKAP to a specific subcellular location. AKAP5 (also known as P75, AKAP75, or AKAP79) is predominantly expressed in neuronal tissues and cells where it serves to localize type II PKA to post-synaptic densities. AKAP5 specifically binds to the regulatory subunit of PKAII beta, anchoring the enzyme to the plasma membrane and sites of cytoskeletal/membrane junctions. The other binding domains of AKAP5 have been shown to interact with calmodulin, PP2B, and calcineurin suggesting that AKAP5 may act to coordinate the cAMP- and Ca2+-sensing pathways in various cell types.
Specifications
| AKAP5 | |
| Polyclonal | |
| Unconjugated | |
| AKAP5 | |
| 3526401B18Rik; A kinase (PRKA) anchor protein 5; AKAP 150; AKAP 5; AKAP 79; AKAP150; AKAP5; AKAP-5; AKAP75; AKAP79; A-kinase anchor protein 150 kDa; A-kinase anchor protein 5; A-kinase anchor protein 79 kDa; A-kinase anchoring protein 5; A-kinase anchoring protein 75/79; BB098886; cAMP-dependent protein kinase regulatory subunit II high affinity-binding protein; Gm258; H21; P150; RII-B-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171026, 238276, 9495 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| D3YVF0, P24587, P24588 | |
| AKAP5 | |
| Synthetic peptide corresponding to the N-terminal of human AKAP 5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction