Learn More
ADAM-like, decysin 1, Mouse, Clone: 6C4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ADAMDEC1.
Brand: Abnova H00027299-M01.100ug
Description
This encoded protein is thought to be a secreted protein belonging to the disintegrin metalloproteinase family. Its expression is upregulated during dendritic cells maturation. This protein may play an important role in dendritic cell function and their interactions with germinal center T cells. [provided by RefSeq
Sequence: PDVPFNTKCPSGSCVMNQYLSSKFPKDFSTSCRAHFERYLLSQKPKCLLQAPIPTNIMTTPVCGNHLLEVGEDCDCGSPKECTNLCCEALTCKLKPGTDCGGDAPNHTTESpecifications
| ADAM-like, decysin 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014479 | |
| ADAMDEC1 | |
| ADAMDEC1 (NP_055294, 361 a.a. ∼ 470 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Signal Transduction | |
| 27299 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 6C4 | |
| Mouse monoclonal antibody raised against a partial recombinant ADAMDEC1. | |
| ADAMDEC1 | |
| M12.219 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Safety and Handling
- ADAMDEC1 monoclonal antibody (M01), clone 6C4
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phisphate monobasic
Your input is important to us. Please complete this form to provide feedback related to the content on this product.