Learn More
ADAM2, Mouse, Clone: 4A2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ADAM2.
Brand: Abnova H00002515-M01.100ug
Description
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions. [provided by RefSeq
Sequence: RLDPFFKQQAVCGNAKLEAGEECDCGTEQDCALIGETCCDIATCRFKAGSNCAEGPCCENCLFMSKERMCRPSFEECDLPEYCNGSSASCPENHYVQTGHSpecifications
| ADAM2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001464 | |
| ADAM2 | |
| ADAM2 (NP_001455, 376 a.a. ∼ 475 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4A2 | |
| Mouse monoclonal antibody raised against a partial recombinant ADAM2. | |
| ADAM2 | |
| CRYN1/CRYN2/FTNB/PH-30b/PH30 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2515 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.