Learn More
anti-active Caspase 3 , Polyclonal, Abcam™
Brand: Abcam AB13847-100UG
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Human, Xenopus laevis, Fruit fly (Drosophila melanogaster), Indian Muntjac, Zebrafish, Rhesus monkey. Predicted to work with Dog, Pig, Chinese Hamster
Immunogen: Synthetic peptide conjugated to KLH derived from within residues 150 - 250 of Human active Caspase 3.
Applications:
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
| active Caspase 3 | |
| 0.9-1mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Cell Biology, Neuroscience | |
| Canine, Drosophila, Hamster, Human, Indian Muntjac, Mouse, Pig, Rat, Rhesus Monkey, Xenopus species, Zebrafish |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide conjugated to KLH derived from within residues 150 - 250 of Human active Caspase 3. | |
| 100 μg | |
| Primary | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.