Learn More
anti-ACOX1, Polyclonal PA127898, Invitrogen™
Specifically detect ACOX1 in Human samples using Thermo Scientifc™ Pierce™ anti-ACOX1 Polyclonal, validated for WB
Brand: Thermo Scientific PA1-27898
Description
Specifically detect ACOX1 in Human samples using Thermo Scientifc™ Pierce™ anti-ACOX1 Polyclonal, validated for WB
Catalyzes the desaturation of very long chain acyl-CoAs to 2-trans-enoyl-CoAs. The protein encoded by this gene is the first enzyme of the fatty acid beta-oxidation pathway. which catalyzes the desaturation of acyl-CoAs to 2-trans-enoyl-CoAs. It donates e
Host Species: Chicken
Cross Reactivity: Human
Isotype: IgY
Immunogen: Synthetic peptide corresponding to human Acyl-coenzyme A oxidase 1. Peroxisomal.
Formulation: 50μg of Antigen affinity chromatography purified antibody in PBS buffer;
PA1-27898 detects ACOX1 in Human samples. PA1-27898 has been successfully used in WB procedures.PA1-27898 The immunogen is Synthetic peptide corresponding to human Acyl-coenzyme A oxidase 1. Peroxisomal.
Specifications
| ACOX1 | |
| Polyclonal | |
| ACOX1 | |
| Synthetic peptide corresponding to human Acyl-coenzyme A oxidase 1. Peroxisomal. | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C. Avoid freeze/Thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgY |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.