missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA112756
This item is not returnable.
View return policy
Description
ASM Polyclonal Antibody for Western Blot, ELISA
The protein encoded by this gene is a lysosomal acid sphingomyelinase that converts sphingomyelin to ceramide. The encoded protein also has phospholipase C activity. Defects in this gene are a cause of Niemann-Pick disease type A (NPA) and Niemann-Pick disease type B (NPB). Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| ASM | |
| Polyclonal | |
| Unconjugated | |
| Smpd1 | |
| Acid sphingomyelinase; ASM; aSMase; A-SMase; NPD; Smpd1; Sphingomyelin phosphodiesterase; sphingomyelin phosphodiesterase 1; sphingomyelin phosphodiesterase 1, acid lysosomal; Zn-SMase | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6609 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.08% sodium azide | |
| P17405 | |
| Smpd1 | |
| Synthetic peptide derived from human acid sphingomyelinase. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction