missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Sensitive Ion Channel 2 Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Brand: Abcam AB5460-50UL
This item is not returnable.
View return policy
Description
Anti-Acid Sensitive Ion Channel 2 Antibody, neuronal is an antibody against Acid Sensitive Ion Channel 2 for use in western blotting.Specifications
| Acid Sensitive Ion Channel 2 | |
| Polyclonal | |
| Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, 5% sucrose and 0.025% sodium azide. Reconstitute with 50μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min). | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Rat |
| Western Blot | |
| Unconjugated | |
| ACCN1; degenerin; BNC1; hBNaC1; MDEG; BNaC1; ACCN; ASIC2a; BNAC1; ASIC2 | |
| Highly purified peptide corresponding to residues 2-18 of rat or human ASIC2 (Accession Q16515). | |
| 50 μL | |
| NM_001094.4; NM_183377.1 | |
| −20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction