missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Sensitive Ion Channel 1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Brand: Abcam AB9034-50UL
This item is not returnable.
View return policy
Description
Anti-Acid Sensitive Ion Channel 1 Antibody detects level of Acid Sensitive Ion Channel 1 and has been published and validated for use in western blotting.Specifications
| Acid Sensitive Ion Channel 1 | |
| Polyclonal | |
| Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Rat |
| Western Blot | |
| Unconjugated | |
| ACCN2; BNaC2; ASIC1A; hBNaC2; ASIC1; ASIC; BNAC2 | |
| Synthetic peptide from rat ASIC1 (Accession P55926).The immunogen sequence is identical in human and 19/20 residues identical in mouse. | |
| 50 μL | |
| NM_001095.2; NM_020039.2 | |
| Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction