missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534855
This item is not returnable.
View return policy
Description
Recommended positive controls: MCF-7. Predicted reactivity: Mouse (100%), Rat (99%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in macrophage lipid homeostasis.
Specifications
| ABCG4 | |
| Polyclonal | |
| Unconjugated | |
| Abcg4 | |
| 6430517O04Rik; ABCG4; ATP binding cassette subfamily G member 4; ATP-binding cassette sub-family G member 4; ATP-binding cassette, subfamily G (WHITE), member 4; ATP-binding cassette, sub-family G (WHITE), member 4; putative ABC transporter; WHITE2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64137 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.72 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9H172 | |
| Abcg4 | |
| Recombinant fragment corresponding to a region within amino acids 1 and 201 of ABCG4 (Uniprot ID#Q9H172) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction