Learn More
ATP-binding cassette, sub-family C (CFTR/MRP), member 6, Mouse, Clone: 5D8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ABCC6.
Brand: Abnova H00000368-M02.100ug
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). The encoded protein, a member of the MRP subfamily, is involved in multi-drug resistance. Mutations in this gene cause pseudoxanthoma elasticum. Alternatively spliced transcript variants that encode different proteins have been described for this gene. [provided by RefSeq
Sequence: IAEMGSYQELLQRKGALVCLLDQARQPGDRGEGETEPGTSTKDPRGTSAGRRPELRRERSIKSVPEKDRTTSEAQTEVPLDDPDRAGWPAGKDSIQYGRVSpecifications
| ATP-binding cassette, sub-family C (CFTR/MRP), member 6 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001171 | |
| ABCC6 | |
| ABCC6 (NP_001162, 831 a.a. ∼ 930 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 368 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Western Blot | |
| 5D8 | |
| Mouse monoclonal antibody raised against a partial recombinant ABCC6. | |
| ABCC6 | |
| ABC34/ARA/EST349056/MLP1/MOATE/MRP6/PXE/PXE1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.