Learn More
ATP-binding cassette, sub-family B (MDR/TAP), member 9, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ABCB9.
Brand: Abnova H00023457-A01.50uL
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. The function of this half-transporter has not yet been determined; however, this protein may play a role in lysosomes. Alternative splicing of this gene results in distinct isoforms which are likely to have different substrate specifications. [provided by RefSeq
Sequence: FIDRQPTMVHDGSLAPDHLEGRVDFENVTFTYRTRPHTQVLQNVSFSLSPGKVTALVGPSGSGKSSCVNILENFYPLEGGRVLLDGKPISAYDHKYLHRSpecifications
| ATP-binding cassette, sub-family B (MDR/TAP), member 9 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ABCB9. | |
| ABCB9 | |
| EST122234/KIAA1520/TAPL | |
| Mouse | |
| 50 μL | |
| Yes | |
| 23457 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_019625 | |
| ABCB9 | |
| ABCB9 (NP_062571, 482 a.a. ∼ 580 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.