missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AAV4 Virus VP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522816
This item is not returnable.
View return policy
Description
AAV4 Virus VP3 Polyclonal Antibody for Western Blot
Adeno-associated virus (AAV) is a small virus which infects humans and some other primate species. AAV is not currently known to cause disease. The virus causes a very mild immune response, lending further support to its apparent lack of pathogenicity.
Specifications
| AAV4 Virus VP3 | |
| Polyclonal | |
| Unconjugated | |
| AAV; AAV-4 VP3; AdV4; AV4; HADV-4; Lentivirus | |
| Directed against a synthetic peptide within residues 450-475 of 81 kDa capsid VP3 protein of AAV 4. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction