Learn More
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant AASDHPPT.
Brand: Abnova H00060496-A01.50uL
Beskrivning
The protein encoded by this gene is similar to Saccharomyces cerevisiae LYS5, which is required for the activation of the alpha-aminoadipate dehydrogenase in the biosynthetic pathway of lysine. Yeast alpha-aminoadipate dehydrogenase converts alpha-biosynthetic-aminoadipate semialdehyde to alpha-aminoadipate. It has been suggested that defects in the human gene result in pipecolic acidemia. [provided by RefSeq
Sequence: MVFPAKRFCLVPSMEGVRWAFSCGTWLPSRAEWLLAVRSIQPEEKERIGQFVFARDAKAAMAGRLMIRKLVAEKLNIPWNHIRLQRTAKGKPVLAKDSSSpecifikationer
| aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant AASDHPPT. | |
| AASDHPPT | |
| AASD-PPT/CGI-80/DKFZp566E2346/LYS2/LYS5 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 60496 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_015423 | |
| AASDHPPT | |
| AASDHPPT (NP_056238, 1 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.