missing translation for 'onlineSavingsMsg'
Learn More

5-Lipoxygenase Mouse, Unlabeled, Clone: 33, BD

Mouse Monoclonal Antibody

£223.06 - £284.36

Specifications

Antigen 5-Lipoxygenase
Clone 33
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15875589
View Documents
BD Biosciences
610694
50 μg
ONLINE EXCLUSIVE PRICE
£233.00
£223.06 / 1 set
Save £9.94
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15885589
View Documents
BD Biosciences
610695
150 μg
ONLINE EXCLUSIVE PRICE
£297.00
£284.36 / 1 set
Save £12.64
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

5-Lipoxygenase (5-LO) is the initial enzyme that converts arachidonic acid to leukotrienes, which are important inflammatory and vasoconstrictive metabolites. It is activated in response to a number of stimuli, such as differentiation and allergen challenges. The 5-LO gene, abundantly expressed in placenta, lung, and leukocytes, encodes a protein of 674 amino acids with an apparent molecular weight of 78kDa. 5-LO is a Ca2+ and ATP-dependent enzyme that translocates from the cytosol to either a nuclear or plasma membrane compartment following activation. A proline-rich domain of 5-LO (amino acids 566-577) has been identified as a binding site for the PTyr-binding protein, Grb2. This Grb2 site links tyrosine kinases with activation and redistribution of 5-LO. Furthermore, tyrosine kinase inhibitors increase the activity of 5-LO and block the enzyme's subcellular redistribution.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

5-Lipoxygenase
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
33
Western Blot
Unconjugated
RUO
Chicken, Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.