missing translation for 'onlineSavingsMsg'
Learn More

Rat alpha 2u-Globulin Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£167.00 - £1795.00

Specifications

Antigen alpha 2u-Globulin
Clone 129736
Dilution Immunohistochemistry 8-25 ug/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18765528
GREENER_CHOICE
View Documents
R&D Systems
MAB586-SP
25 μg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18746958
GREENER_CHOICE
View Documents
R&D Systems
MAB586-100
100 μg
£453.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18716958
GREENER_CHOICE
View Documents
R&D Systems
MAB586-500
500 μg
£1795.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

alpha 2u-Globulin Monoclonal specifically detects alpha 2u-Globulin in Rat samples. It is validated for Immunohistochemistry.
Specifications

Specifications

alpha 2u-Globulin
Immunohistochemistry 8-25 ug/mL
Monoclonal
Mouse
Rat
AUG, Mup, PGCL1
Rat urine-derived alpha 2u -Globulin
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects rat alpha 2μ -Globulin in direct ELISAs.
129736
Immunohistochemistry
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
259246
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.