Learn More
β2-Microglobulin Mouse anti-Human, FITC, Clone: TÜ99, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551338
Description
β 2-microglobulin is the 12 kDa β chain which noncovalently associates with the 44 kDa α chain to form the HLA Class I antigen complex. This antibody reacts with free or complexed β 2-microglobulin. TÜ99 does not react with β 2-microglobulin of other species. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections using TUF pretreatment.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| β2-Microglobulin | |
| Monoclonal | |
| FITC | |
| Mouse | |
| 100 Tests | |
| Cell Biology | |
| Human | |
| IgM κ |
| Flow Cytometry | |
| TÜ99 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.