Learn More
β2-Microglobulin Mouse anti-Human, Unlabeled, Clone: TÜ99, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555550
Description
β 2-microglobulin is the 12 kDa β chain which noncovalently associates with the 44 kDa α chain to form the HLA Class I antigen complex. This antibody reacts with free or complexed β 2-microglobulin. TÜ99 does not react with β 2-microglobulin of other species. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections using TUF pretreatment.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| β2-Microglobulin | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| TÜ99 | |
| Unconjugated | |
| Mouse | |
| 0.1 mg | |
| Cell Biology | |
| Human | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.