missing translation for 'onlineSavingsMsg'
Learn More

Anthrone, ACS

Product Code. 11357338
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357338 100 g 100g
11367338 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357338 Supplier Thermo Scientific Alfa Aesar Supplier No. 030739.22
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for determination of carbohydrates | CAS: 90-44-8 | C14H10O | 194.23 g/mol

Used for determination of carbohydrates Anthrone is used for the determination of carbohydrates. It is used as a general reagent in the colorimetric determination of carbohydrates in yeast and plant extracts. It is involved in organic synthesis. It is utilized for the determination of sugar in body fluids. It finds application as a laxative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-44-8
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001187
InChI Key RJGDLRCDCYRQOQ-UHFFFAOYSA-N
Synonym anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744
PubChem CID 7018
ChEBI CHEBI:33835
IUPAC Name 10H-anthracen-9-one
SMILES O=C1C2=CC=CC=C2CC2=CC=CC=C12

Specifications

Melting Point 155°C to 158°C
Quantity 100 g
Beilstein 1910173
Sensitivity Light sensitive
Merck Index 14,691
Solubility Information Soluble in acetone,hot benzene,concentrated sulfuric acid,hot dilute alkali and most organic solvents. Insoluble in water.
Formula Weight 194.23
Grade ACS
Physical Form Crystalline
Chemical Name or Material Anthrone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.