missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anthrax Protective Antigen Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141128
This item is not returnable.
View return policy
Description
Suggested positive control: PA recombinant protein.
Anthrax infection is initiated by the inhalation, ingestion, or cutaneous contact with Bacillus anthracis endospores. B. anthracis produces three polypeptides that comprise the anthrax toxin: protective antigen (PA), lethal factor (LF), and edema factor (EF). PA binds to two related proteins on the cell surface; these are termed tumor epithelial marker 8 (TEM8)/anthrax toxin receptor (ATR) and capillary morphogenesis protein 2 (CMG2), although it is still unclear which is physiologically relevant. Following PA binding to its receptor, PA is cleaved into two fragments by a furin-like protease. The bound fragment binds both LF and EF; the resulting complex is then endocytosed which allows the translocation of LF and EF into the cytoplasm. These toxins are usually sufficient to cause rapid cell death, and often the death of the organism.
Specifications
| Anthrax Protective Antigen | |
| Polyclonal | |
| Unconjugated | |
| Anthrax PA | |
| Synthetic peptides corresponding to amino acids 186-192 of the Anthrax PA protein (approx. 83 kDa). | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| Western Blot | |
| Conc. Not Determined | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction