missing translation for 'onlineSavingsMsg'
Learn More

Anthrax PA Antibody (PA6), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18420528 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420528 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420528 Supplier Novus Biologicals Supplier No. NBP197535JF646

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Anthrax PA Monoclonal antibody specifically detects Anthrax PA in Bacteria samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Anthrax PA
Applications Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PA6
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias anthrax PA, Anthrax toxins translocating protein, Bacillus anthracis protective antigen, PA, PA 83, PA83, PAG, Paga, Protective antigen
Host Species Mouse
Immunogen Anthrax protective antigen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.