missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anthrax PA Antibody (PA6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-97535CL1
This item is not returnable.
View return policy
Description
Anthrax PA Monoclonal antibody specifically detects Anthrax PA in Bacteria samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| Anthrax PA | |
| Monoclonal | |
| CoraFluor 1 | |
| anthrax PA, Anthrax toxins translocating protein, Bacillus anthracis protective antigen, PA, PA 83, PA83, PAG, Paga, Protective antigen | |
| Anthrax protective antigen. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA, Immunoprecipitation, Western Blot | |
| PA6 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction