missing translation for 'onlineSavingsMsg'
Learn More

Anthraquinone-2-carboxylic acid, 98%

Product Code. 15467428
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467428 5 g 5g
15457428 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467428 Supplier Thermo Scientific Alfa Aesar Supplier No. H60929.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 117-78-2 | C15H8O4 | 252.225 g/mol

Anthraquinone 2-carboxylic acid (AQ) as a novel electron shuttling mediator and attached electron relay for HRP. Efficient method for the generation of hydrogen peroxide by aerobic photooxidation of 2-propanol using anthraquinone-2-carboxylic acid and molecular oxygen is investigated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthraquinone 2-carboxylic acid (AQ) as a novel electron shuttling mediator and attached electron relay for HRP. Efficient method for the generation of hydrogen peroxide by aerobic photooxidation of 2-propanol using anthraquinone-2-carboxylic acid and molecular oxygen is investigated.

Solubility
Soluble in hot Acetic acid(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-78-2
Molecular Formula C15H8O4
Molecular Weight (g/mol) 252.225
MDL Number MFCD00001231
InChI Key ASDLSKCKYGVMAI-UHFFFAOYSA-N
Synonym anthraquinone-2-carboxylic acid, 9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, 2-carboxyanthraquinone, 2-anthraquinonecarboxylic acid, 2-carboxyl-9,10-anthraquinone, anthraquinone-2-carboxylate, 9,10-dihydro-9,10-dioxo-2-anthracenecarboxylic acid, 9,10-anthraquinone 2-carboxylic acid, .beta.-anthraquinonecarboxylic acid, anthraquinone-.beta.-carboxylic acid
PubChem CID 67030
IUPAC Name 9,10-dioxoanthracene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)C(=O)O

Specifications

Melting Point 287°C to 289°C
Quantity 5 g
Solubility Information Soluble in hot Acetic acid(almost transparency).
Formula Weight 252.22
Percent Purity 98%
Chemical Name or Material Anthraquinone-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.