missing translation for 'onlineSavingsMsg'
Learn More

Anthranilic acid, 98+%

Product Code. 13448298 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
5 kg
Unit Size:
100g
5kg
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Product Code. 13448298

Brand: Thermo Scientific Acros 104910051

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

CAS: 118-92-3 | C7;H7;NO2; | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-92-3
Molecular Formula C7;H7;NO2;
Molecular Weight (g/mol) 137.14

Specifications

Melting Point 144.0°C to 148.0°C
Assay Percent Range 98+%
Packaging Plastic Drum
Beilstein 14, 310
Merck Index 15, 417
Solubility Information Solubility aq. soln.: 5.7 g/L (25°C). Other solubilities: readily soluble in ethanol, esters, acetone, soluble in pyridine, hot chloroform, hot benzene
Density 1.4100 g/mL
Quantity 5 kg
Flash Point 150°C
Formula Weight 137.14
Specific Gravity 1.4100
Chemical Name or Material Anthranilic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.