missing translation for 'onlineSavingsMsg'
Learn More

Anthranilic acid, 98+%

Product Code. 10182700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
5 kg
Unit Size:
100g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10182700 100 g 100g
13448298 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 10182700 Supplier Thermo Scientific Acros Supplier No. 104911000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

CAS: 118-92-3 | C7;H7;NO2; | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-92-3
Molecular Formula C7;H7;NO2;
Molecular Weight (g/mol) 137.14

Specifications

Melting Point 144.0°C to 148.0°C
Assay Percent Range 98+%
Packaging Plastic Bottle
Beilstein 14, 310
Merck Index 15, 417
Solubility Information Solubility aq. soln.: 5.7 g/L (25°C). Other solubilities: readily soluble in ethanol, esters, acetone, soluble in pyridine, hot chloroform, hot benzene
Density 1.4100 g/mL
Quantity 100 g
Flash Point 150°C
Formula Weight 137.14
Specific Gravity 1.4100
Chemical Name or Material Anthranilic acid
Show More Show Less
compliance-icons
Product Identifier
  • Anthranilic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.