Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Anthranillic acid is an organic aromatic acid consisting of benzoic acid with an o-substituted amino group
- Produced as a metabolite in the L-Tryptophan-kynurenine pathway, anthranilic acid is found in both mice and humans
- Esters of anthranilic acid are used in perfumes, dyes, synthetic flavor development, and as bird and insect repellents
- Anthranilic acid has been used as a fungicide/mold inhibitor
Applications
- Anthranilic acid is an organic reagent used in preparation of dyes, pigments, and perfumes
- It has been used to fluorescently tag reducing glycans for analysis by HPLC or LC/MS
- It can be used as a fluorophore group in FRET applications
Specifications
Specifications
| CAS | 118-92-3 |
| Molecular Formula | C7H7NO2 |
| Quantity | 500 g |
| InChI Key | RWZYAGGXGHYGMB-UHFFFAOYSA-N |
| SMILES | NC1=CC=CC=C1C(O)=O |
| IUPAC Name | 2-aminobenzoic acid |
| Molecular Weight (g/mol) | 137.14 |
| Chemical Name or Material | Anthranilic acid, 98+% |
Safety and Handling
- Anthranilic acid
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.